Thermo Scientific Chemicals

Chloramine-T trihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals

Catalog number: 042374.22
100 g, Each
Thermo Scientific Chemicals

Chloramine-T trihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals

Catalog number: 042374.22
100 g, Each
Quantity
Catalog number: 042374.22
also known as 042374-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7080-50-4
IUPAC Name
sodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
Molecular Formula
C7H13ClNNaO5S
InChI Key
RNROOTWAJBXJBD-UHFFFAOYSA-M
SMILES
O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl
Comment
Suitable for determination of bromide P.T.
Comment
Clarity of aqueous solution P.T.
Impurity content
Insoluble in alcohol 1.5%
pH
of a 5% solution 8.0-10.0 at25?

Description

Reagent for selective oxidation of methionineChloramine-T trihydrate is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals. It combines with iodogen or lactoperoxidase and is commonly used for labeling peptides and proteins with radioiodine isotopes. Hypochlorite released from chloramine-T acts as an effective oxidizing agent for iodide to form iodine monochloride (ICl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text