Poly(2-ethyl-2-oxazoline), M.W. 500,000, Thermo Scientific Chemicals
Poly(2-ethyl-2-oxazoline), M.W. 500,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Poly(2-ethyl-2-oxazoline), M.W. 500,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 042398.22
also known as 042398-22
Price (USD)/ Each
36.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
36.00
Add to cart
Poly(2-ethyl-2-oxazoline), M.W. 500,000, Thermo Scientific Chemicals
Catalog number042398.22
Price (USD)/ Each
36.00
-
Add to cart
Chemical Identifiers
CAS25805-17-8
Molecular Formula(C5H9NO)n
SMILESCCC(=O)N(-*)CC-*
MDL NumberMFCD00134377
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
CommentPolydispersity range 3-4
CommentTarget molecualr weight 500,000
Loss on Drying≤1.0% (2 hours, 125?C)
Viscosity60-80 cSt (kinematic, 10%, in water, @ 100?F / 37.8?C)
View more
Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only