Thermo Scientific Chemicals

N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol, Thermo Scientific Chemicals

Catalog number: 042405.A1
1 kg, Each
Thermo Scientific Chemicals

N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol, Thermo Scientific Chemicals

Catalog number: 042405.A1
1 kg, Each
Quantity
Catalog number: 042405.A1
also known as 042405-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2680-03-7
IUPAC Name
N,N-dimethylprop-2-enamide
Molecular Formula
C5H9NO
InChI Key
YLGYACDQVQQZSW-UHFFFAOYSA-N
SMILES
CN(C)C(=O)C=C
Form
Liquid
Assay from Suppliers CofA
≥97.5%
Refractive Index
1.4650-1.4750
Stabilizer
50-150 ppm MEHQ
Appearance (Color)
Clear colorless

Description

N,N-Dimethylacrylamide is a useful monomer for synthetic fibers and plastics. It is also used in the synthesis of photoresponsive polymers and copolymers by grafting polysiloxane on the side chain of DMA by homopolymerization or grafting cellulose to form cellulose polymers and copolymerization of methyl methacrylate with DMA or copolymerization of DMA with styrene and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text