Thermo Scientific Chemicals

Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042408.09
10 g, Each
Thermo Scientific Chemicals

Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic), Thermo Scientific Chemicals

Catalog number: 042408.09
10 g, Each
Quantity
Catalog number: 042408.09
also known as 042408-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13536-59-9
IUPAC Name
proton bromide
Molecular Formula
BrH
InChI Key
CPELXLSAUQHCOX-FTGQXOHASA-N
SMILES
[1H+].[Br-]
Assay from Supplier's CofA
≥99.0% Isotopic enrichment by ¹H nmr
Assay from Supplier's CofA
≥98.0% ¹H nmr

Description

NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text