Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic), Thermo Scientific Chemicals
Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic), Thermo Scientific Chemicals

NMR Solvent
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042408.18
also known as 042408-18
Price (USD)/ Each
238.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
281.00 
Save 42.35 (15%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
238.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
281.00 
Save 42.35 (15%)
Add to cart
Deuterium bromide, 48% w/w in D{2}O, 99+% (Isotopic), Thermo Scientific Chemicals
Catalog number042408.18
Price (USD)/ Each
238.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
281.00 
Save 42.35 (15%)
-
Add to cart
Chemical Identifiers
CAS13536-59-9
IUPAC Nameproton bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-FTGQXOHASA-N
SMILES[1H+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.0% Isotopic enrichment by ¹H nmr
Assay from Supplier's CofA≥98.0% ¹H nmr
NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only