Thermo Scientific Chemicals

Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical), Thermo Scientific Chemicals

Catalog number: 042410.22
100 g, Each
Thermo Scientific Chemicals

Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical), Thermo Scientific Chemicals

Catalog number: 042410.22
100 g, Each
Quantity
Catalog number: 042410.22
also known as 042410-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40372-72-3
IUPAC Name
4,4,15,15-tetraethoxy-3,16-dioxa-8,9,10,11-tetrathia-4,15-disilaoctadecane
Molecular Formula
C18H42O6S4Si2
InChI Key
VTHOKNTVYKTUPI-UHFFFAOYSA-N
SMILES
CCO[Si](CCCSSSSCCC[Si](OCC)(OCC)OCC)(OCC)OCC
Appearance (Color)
Clear yellow to brown
Form
Liquid
Assay from Suppliers CofA
Sulfur, S: ≥22.0%

Description

Bis[3-(triethoxysilyl)propyl]tetrasulfide is used in radial tires, rubber tire treads, carcass, tire walls, solid rubber tires and other rubber products. It acts as a chemical intermediate and as a cross linking agent. Furthermore, it acts as a coupling agent used to chemically modify silica surface. In addition, it serves as a curing agent for good heat aging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text