Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 042412.30
also known as 042412-30
Price (USD)/ Each
66.65
Online exclusive
73.50 
Save 6.85 (9%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
66.65
Online exclusive
73.50 
Save 6.85 (9%)
Add to cart
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Catalog number042412.30
Price (USD)/ Each
66.65
Online exclusive
73.50 
Save 6.85 (9%)
-
Add to cart
Chemical Identifiers
CAS541-02-6
IUPAC Namedecamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
Molecular FormulaC10H30O5Si5
InChI KeyXMSXQFUHVRWGNA-UHFFFAOYSA-N
SMILESC[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormClear liquid
Assay (unspecified)> 96.0%
Decamethylcyclopentasiloxane is employed in cosmetics like deodorants, sunblocks, hair sprays and personal care products. Furthermore, it is used in silicone based personal lubricants. In addition, it is a cyclic volatile methylsiloxane used in hair conditioners, thereby brushing the hair without breakage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only