Thermo Scientific Chemicals

(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 042413.AD
50 mL, Each
Thermo Scientific Chemicals

(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 042413.AD
50 mL, Each
Quantity
Catalog number: 042413.AD
also known as 042413-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2530-87-2
IUPAC Name
(3-chloropropyl)trimethoxysilane
Molecular Formula
C6H15ClO3Si
InChI Key
OXYZDRAJMHGSMW-UHFFFAOYSA-N
SMILES
CO[Si](CCCCl)(OC)OC
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay from Suppliers CofA
≥97%

Description

(3-Chloropropyl)trimethoxysilane is used as adhesives and sealant chemicals. It is also used in the preparation of 3-iodopropyltrimethoxysilane by reacting with sodium iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text