Thermo Scientific Chemicals
Chlorodimethyl-n-octadecylsilane, 95%, Thermo Scientific Chemicals
Catalog number: 042414.36
500 g, Each
Thermo Scientific Chemicals
Chlorodimethyl-n-octadecylsilane, 95%, Thermo Scientific Chemicals
Catalog number: 042414.36
500 g, Each
Quantity
Catalog number: 042414.36
also known as 042414-36
Price (USD)
Quantity
-
Have Questions?
Description
Chlorodimethyl-n-octadecylsilane is used for preparation of hydrophobic surfaces and chain terminator.