Buffer solution, pH 5.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Buffer solution, pH 5.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer solution, pH 5.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 042417.AP
also known as 042417-AP
Price (USD)/ Each
30.30
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
30.30
Add to cart
Buffer solution, pH 5.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Catalog number042417.AP
Price (USD)/ Each
30.30
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialBuffer solution, pH 5.00 (±0.01 at 25°C)
Name NoteNIST Traceable
Quantity500 mL
Recommended StorageAmbient temperatures
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only