Buffer solution, pH 6.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Buffer solution, pH 6.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer solution, pH 6.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 042418.AP
also known as 042418-AP
Price (USD)/ Each
30.70
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
30.70
Add to cart
Buffer solution, pH 6.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Catalog number042418.AP
Price (USD)/ Each
30.70
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialBuffer solution, pH 6.00 (±0.01 at 25°C)
Name NoteNIST Traceable
Health Hazard 1H290-H315-H319
Quantity500 mL
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
View more
Buffer solution (pH 6) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only