Buffer solution, pH 9.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Buffer solution, pH 9.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer solution, pH 9.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
500 mL
1 L
Catalog number 042421.K7
also known as 042421-K7
Price (USD)/ Each
70.65
Online exclusive
78.00 
Save 7.35 (9%)
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
70.65
Online exclusive
78.00 
Save 7.35 (9%)
Add to cart
Buffer solution, pH 9.00 (+/-0.024 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Catalog number042421.K7
Price (USD)/ Each
70.65
Online exclusive
78.00 
Save 7.35 (9%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialBuffer solution, pH 9.00 (±0.01 at 25°C)
Name NoteNIST Traceable
Quantity4 L
Recommended StorageAmbient temperatures
TSCAYes
View more
Buffer solution (pH 9) is used as a means of keeping the pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing. It is also used in chemical analysis and in calibration of pH meters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only