Buffer solution, pH 12.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Buffer solution, pH 12.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer solution, pH 12.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 042425.AP
Price (USD)/ Each
28.65
Online exclusive
32.10 
Save 3.45 (11%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
28.65
Online exclusive
32.10 
Save 3.45 (11%)
Add to cart
Buffer solution, pH 12.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals
Catalog number042425.AP
Price (USD)/ Each
28.65
Online exclusive
32.10 
Save 3.45 (11%)
-
Add to cart
Chemical Identifiers
CAS1336-21-6
SpecificationsSpecification SheetSpecification Sheet
pH11.99-12.01 at 25 deg C
Appearance (Color)Colorless
FormLiquid
Buffer solution (pH 12.00) is used as a means of keeping the pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only