(3-Mercaptopropyl)methyldimethoxysilane, 95%, Thermo Scientific Chemicals
(3-Mercaptopropyl)methyldimethoxysilane, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(3-Mercaptopropyl)methyldimethoxysilane, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Catalog number 042426.AK
also known as 042426-AK
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
Add to cart
(3-Mercaptopropyl)methyldimethoxysilane, 95%, Thermo Scientific Chemicals
Catalog number042426.AK
Price (USD)/ Each
197.65
Online exclusive
219.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS31001-77-1
IUPAC Name3-[(dimethoxymethyl)silyl]propane-1-thiol
Molecular FormulaC6H16O2SSi
InChI KeyLOOUJXUUGIUEBC-UHFFFAOYSA-N
SMILESCOC(OC)[SiH2]CCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay from Suppliers CofA≥94%
(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only