Cadmium, Oil based standard solution, Specpure™ Cd 5000μg/g, Thermo Scientific Chemicals
Cadmium, Oil based standard solution, Specpure™ Cd 5000μg/g, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium, Oil based standard solution, Specpure™ Cd 5000μg/g, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042436.18
also known as 042436-18
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.30 
Save 10.65 (15%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.30 
Save 10.65 (15%)
Add to cart
Cadmium, Oil based standard solution, Specpure™ Cd 5000μg/g, Thermo Scientific Chemicals
Catalog number042436.18
Price (USD)/ Each
60.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
71.30 
Save 10.65 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialCadmium, Oil based standard solution
Name NoteCd 5000μg/g
Health Hazard 1H350
Quantity50 g
Health Hazard 2GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only