Thermo Scientific Chemicals

Acetone-d{6}, 99.9% (Isotopic), contains 1% v/v TMS, Thermo Scientific Chemicals

Catalog number: 042458.14
25 g, Each
Thermo Scientific Chemicals

Acetone-d{6}, 99.9% (Isotopic), contains 1% v/v TMS, Thermo Scientific Chemicals

Catalog number: 042458.14
25 g, Each
Quantity
Catalog number: 042458.14
also known as 042458-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
666-52-4
IUPAC Name
(²H₆)propan-2-one
Molecular Formula
C3H6O
InChI Key
CSCPPACGZOOCGX-WFGJKAKNSA-N
SMILES
[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥99.5%
Assay from Supplier's CofA
0.8-1.2 v/v TMS
Assay from Supplier's CofA
≥99.5% ¹H nmr

Description

Acetone-d{6} is used to produce Perdeuterio-aceton-cyanhydrin at temperature of 15 - 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text