Thermo Scientific Chemicals

Hafnium(IV) 2,4-pentanedionate, 97%, Thermo Scientific Chemicals

Catalog number: 042463.03
1 g, Each
Thermo Scientific Chemicals

Hafnium(IV) 2,4-pentanedionate, 97%, Thermo Scientific Chemicals

Catalog number: 042463.03
1 g, Each
Quantity
Catalog number: 042463.03
also known as 042463-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17475-67-1
IUPAC Name
hafnium(4+) tetrakis(2,4-dioxopentan-3-ide)
Molecular Formula
C20H28HfO8
InChI Key
GXNMJQUZCICHBX-UHFFFAOYSA-N
SMILES
[Hf+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Appearance (Color)
White to pale cream
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
Hf: ≥99.5% (metals basis)
Elemental Analysis
≥96%
Elemental Analysis
C: 40.7- 42.8%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text