Silver foil, 0.025mm (0.001in) thick, annealed, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals
Silver foil, 0.025mm (0.001in) thick, annealed, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver foil, 0.025mm (0.001in) thick, annealed, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals

Form: approx. 0.16g/25x25mm; Temper: soft
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
50 x 250 mm
Catalog number 042472.FF
also known as 042472-FF
Price (USD)/ Each
53.60
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
53.60
Add to cart
Silver foil, 0.025mm (0.001in) thick, annealed, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number042472.FF
Price (USD)/ Each
53.60
-
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
Name NoteAnnealed
CAS7440-22-4
AppearanceSilver-colored foil
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only