Thermo Scientific Chemicals

7-Methylquinoline, 97%, Thermo Scientific Chemicals

Catalog number: 042475.01
0.25 g, Each
Thermo Scientific Chemicals

7-Methylquinoline, 97%, Thermo Scientific Chemicals

Catalog number: 042475.01
0.25 g, Each
Quantity
Catalog number: 042475.01
also known as 042475-01
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
612-60-2
IUPAC Name
7-methylquinoline
Molecular Formula
C10H9N
InChI Key
KDYVCOSVYOSHOL-UHFFFAOYSA-N
SMILES
CC1=CC=C2C=CC=NC2=C1
Appearance (Color)
Pale yellow
Form
Low melting solid / liquid
Assay (GC)
≥96.0%
Proton NMR
Conforms
Melting Point
34-37?C

Description

7-methylquinoline has been bioassayed as tumor initiator on the skin of Sencar mice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text