Stainless Steel flake, -150 mesh, 1.2 micron thick, Type 316-L, Thermo Scientific Chemicals
Stainless Steel flake, -150 mesh, 1.2 micron thick, Type 316-L, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stainless Steel flake, -150 mesh, 1.2 micron thick, Type 316-L, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 042486.A1
also known as 042486-A1
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
Stainless Steel flake, -150 mesh, 1.2 micron thick, Type 316-L, Thermo Scientific Chemicals
Catalog number042486.A1
Price (USD)/ Each
139.00
-
Add to cart
Specifications
Chemical Name or MaterialStainless Steel flake, Type 316-L
Name Note1.2 micron thick
Recommended StorageAmbient temperatures
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
TSCAYes
View more
Stainless Steel is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.