Thermo Scientific Chemicals

Inhibitor Removal Resin

Catalog number: 042489.14
25 g, Each
Thermo Scientific Chemicals

Inhibitor Removal Resin

Catalog number: 042489.14
25 g, Each
Quantity
Catalog number: 042489.14
also known as 042489-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-70-7
MDL Number
9003-70-7
Assay (special)
Total Volume Capacity: ≥1.15 eq/l
Assay (special)
Moisture Retention Capacity: 53-58%
Form
Powder or crystals or crystalline powder

Description

Inhibitor removal resin is used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol. It is also take part in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text