Zirconium(IV) oxide, cement, Thermo Scientific Chemicals
Zirconium(IV) oxide, cement, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) oxide, cement, Thermo Scientific Chemicals

One component adhesive. Bonds to Zirconium oxide and Silicon. Exhibit good dielectric properties
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042493.30
also known as 042493-30
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZirconium(IV) oxide
CAS1314-23-4
ColorBrown
Health Hazard 1H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only