Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1250, Thermo Scientific Chemicals
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1250, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1250, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
250 mL
Catalog number 042498.K2
also known as 042498-K2
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1250, Thermo Scientific Chemicals
Catalog number042498.K2
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Viscosity9-11 cSt at 25?
FormViscous liquid
Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppress the formation of foams and as a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only