Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000, Thermo Scientific Chemicals
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
250 mL
Catalog number 042504.K2
also known as 042504-K2
Price (USD)/ Each
212.00
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
212.00
Add to cart
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000, Thermo Scientific Chemicals
Catalog number042504.K2
Price (USD)/ Each
212.00
-
Add to cart
Chemical Identifiers
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormViscous liquid
Infrared spectrumConforms
Viscosity54000-66000 cSt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only