Dihydrogen hexabromoosmate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Dihydrogen hexabromoosmate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dihydrogen hexabromoosmate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 042527.06
also known as 042527-06
Price (USD)/ Each
669.65
Online exclusive
744.00 
Save 74.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
669.65
Online exclusive
744.00 
Save 74.35 (10%)
Add to cart
Dihydrogen hexabromoosmate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number042527.06
Price (USD)/ Each
669.65
Online exclusive
744.00 
Save 74.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialDihydrogen hexabromoosmate (IV) hydrate
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
Melting Point(decomposition)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only