Thermo Scientific Chemicals

Gallium(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 042528.22
100 g, Each
Thermo Scientific Chemicals

Gallium(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 042528.22
100 g, Each
Quantity
Catalog number: 042528.22
also known as 042528-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12024-21-4
IUPAC Name
digallium(3+) trioxidandiide
Molecular Formula
Ga2O3
InChI Key
AJNVQOSZGJRYEI-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Ga+3].[Ga+3]
Assay (special)
?2.5 % of Theoretical value (Ga)
Total Metal Impurities
≤0.001% (10 ppm)
Form
Powder
Appearance (Color)
White

Description

Gallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. Used as an evaporated material and sputtering target of 99.999% in dielectric films. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text