Apatite, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals
Apatite, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Apatite, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 042535.06
also known as 042535-06
Price (USD)/ Each
92.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
92.80
Add to cart
Apatite, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals
Catalog number042535.06
Price (USD)/ Each
92.80
-
Add to cart
Chemical Identifiers
CAS1306-05-4
IUPAC Namepentacalcium fluoride triphosphate
Molecular FormulaCa5FO12P3
InChI KeyVSIIXMUUUJUKCM-UHFFFAOYSA-D
SMILES[F-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentApatite is naturally occurring Mineral mined and crushed to approximately 0.06-0.19in without any chemical processing.
Apatite is one of a few minerals produced and used by biological micro-environmental systems. The primary use of apatite is in the manufacture of fertilizer, it is a source of phosphorus. It is occasionally used as a gemstone. Green and blue varieties in finely divided form, are pigments with excellent covering power.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only