Thermo Scientific Chemicals

Galena, naturally occurring mineral, approximately 1-2in, Thermo Scientific Chemicals

Catalog number: 042546.KF
1 Ea., Each
Thermo Scientific Chemicals

Galena, naturally occurring mineral, approximately 1-2in, Thermo Scientific Chemicals

Catalog number: 042546.KF
1 Ea., Each
Quantity
Catalog number: 042546.KF
also known as 042546-KF
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-87-0
IUPAC Name
λ²-lead(2+) sulfanediide
Molecular Formula
PbS
InChI Key
FILXVLQXZYXBSJ-UHFFFAOYSA-N
SMILES
[S--].[Pb++]
Comment
Galena is naturally occurring Lead(II) sulfide mined and crushed to approximately 1-2in without any chemical processing.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text