Glassy carbon spherical powder, 200-400 micron, type 2, Thermo Scientific Chemicals
Glassy carbon spherical powder, 200-400 micron, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon spherical powder, 200-400 micron, type 2, Thermo Scientific Chemicals

Used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 042550.18
also known as 042550-18
Price (USD)/ Each
127.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
127.00
Add to cart
Glassy carbon spherical powder, 200-400 micron, type 2, Thermo Scientific Chemicals
Catalog number042550.18
Price (USD)/ Each
127.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGlassy carbon spherical powder
Health Hazard 1H228
Health Hazard 3P210-P240-P241-P280-P370+P378q
Recommended StorageAmbient temperatures
DOT InformationHazard Class: 4.1; Packaging Group: III
View more

Glassy carbon spherical powder is used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only