Thermo Scientific Chemicals

Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 042562.AP
500 mL, Each
Thermo Scientific Chemicals

Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL, Thermo Scientific Chemicals

Catalog number: 042562.AP
500 mL, Each
Quantity
Catalog number: 042562.AP
also known as 042562-AP
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000 ? 5mg/L
Concentration
1,000mg/L ?1%
Matrix
H₂O
Starting Material
KHC₈H₄O₄

Description

Total Organic Carbon (TOC), is used as a standard solution during the measurement of chemical analysis such as detecting contaminants in drinking water and cooling water. It is also used for analysis of water to be used for semiconductor manufacturing and pharmaceutical use.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text