Aluminum oxide, Alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals
Aluminum oxide, Alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, Alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals

S.A. 6-8m2/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042572.22
also known as 042572-22
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
43.10 
Save 6.45 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
43.10 
Save 6.45 (15%)
Add to cart
Aluminum oxide, Alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number042572.22
Price (USD)/ Each
36.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
43.10 
Save 6.45 (15%)
-
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
CommentCrystal structure: Alpha phase
Particle SizeAPS, D₅₀: typically 0.5-0.7µm
Total Metal Impurities≤0.1% (≤1,000ppm)
View more
Aluminum oxide is widely used in a variety of plastics, rubber, ceramics, refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire, yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamp as far-infrared emission and thermal insulation materials. It is useful for the C single crystal detectors. It plays an important role for the hydrogenation of aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only