Thermo Scientific Chemicals

Silver(I) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 042577.14
25 g, Each
Thermo Scientific Chemicals

Silver(I) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 042577.14
25 g, Each
Quantity
Catalog number: 042577.14
also known as 042577-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20667-12-3
IUPAC Name
(argentiooxy)silver
Molecular Formula
Ag2O
InChI Key
KHJDQHIZCZTCAE-UHFFFAOYSA-N
SMILES
[Ag]O[Ag]
Total Metal Impurities
0.01% max.

Description

Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent, carbon dioxide scrubber, and chemical sensors. It is used in the preparation of other silver compounds, and silver-oxide batteries. In organic chemistry, silver oxide finds use as an oxidizing agent for aldehyes to produce carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text