Thermo Scientific Chemicals

Polyaniline, emeraldine base, Thermo Scientific Chemicals

Catalog number: 042581.14
25 g, Each
Thermo Scientific Chemicals

Polyaniline, emeraldine base, Thermo Scientific Chemicals

Catalog number: 042581.14
25 g, Each
Quantity
Catalog number: 042581.14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
25233-30-1
Molecular Formula
(C12H10N2)x(C12H8N2)y
SMILES
*-N=C1C=CC(C=C1)=NC1=CC=C(NC2=CC=C(NC3=CC=C(-*)C=C3)C=C2)C=C1
MDL Number
MFCD00241446
Form
powder
Melting Point (DSC)
> 200 deg C ( decomposes)
Appearance (Color)
Dark brown
Loss on Drying
< 10 wt % @ 150 deg C by TGA

Description

Conductive polymerPolyaniline is used in chemical vapor sensors, supercapacitors and biosensors. It is also useful for manufacture of electrical conducting yarns, antistatic coatings, electromagnetic shielding, flexible electrodes and printed circuit board. Further, it is used in antistatic and ESD coatings and corrosion protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text