Thermo Scientific Chemicals

Abietic acid, 90+%, Thermo Scientific Chemicals

Catalog number: 042582.03
1 g, Each
Thermo Scientific Chemicals

Abietic acid, 90+%, Thermo Scientific Chemicals

Catalog number: 042582.03
1 g, Each
Quantity
Catalog number: 042582.03
also known as 042582-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
514-10-3
IUPAC Name
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
Molecular Formula
C20H30O2
InChI Key
RSWGJHLUYNHPMX-ONCXSQPRSA-N
SMILES
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1
Assay (HPLC)
> 90.0%
Appearance (Color)
Off-white to pale yellow
Transition Temperature
powder

Description

Abietic acid is used to make ester gums in lacquers, varnishes and soaps. It is also used in metal resonates and plastics. Further, it is utilized in musical instruments to make them less slippery. It acts as a precursor to natural product synthesis. In addition to this, it is used to depackage integrated circuits from their epoxy coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text