Abietic acid, 90+%, Thermo Scientific Chemicals
Abietic acid, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Abietic acid, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 042582.MD
also known as 042582-MD
Price (USD)/ Each
90.65
Online exclusive
100.00 
Save 9.35 (9%)
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
90.65
Online exclusive
100.00 
Save 9.35 (9%)
Add to cart
Abietic acid, 90+%, Thermo Scientific Chemicals
Catalog number042582.MD
Price (USD)/ Each
90.65
Online exclusive
100.00 
Save 9.35 (9%)
-
Add to cart
Chemical Identifiers
CAS514-10-3
IUPAC Name(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
Molecular FormulaC20H30O2
InChI KeyRSWGJHLUYNHPMX-ONCXSQPRSA-N
SMILESCC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)> 90.0%
Appearance (Color)Off-white to pale yellow
Transition Temperaturepowder
Abietic acid is used to make ester gums in lacquers, varnishes and soaps. It is also used in metal resonates and plastics. Further, it is utilized in musical instruments to make them less slippery. It acts as a precursor to natural product synthesis. In addition to this, it is used to depackage integrated circuits from their epoxy coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only