Thermo Scientific Chemicals

Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals

Catalog number: 042583.14
25 g, Each
Thermo Scientific Chemicals

Zirconium(IV) tert-butoxide, 97+%, Thermo Scientific Chemicals

Catalog number: 042583.14
25 g, Each
Quantity
Catalog number: 042583.14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2081-12-1
IUPAC Name
tetrakis(2-methylpropan-2-ol) zirconium
Molecular Formula
C16H40O4Zr
InChI Key
WRMYASGYMABHCC-UHFFFAOYSA-N
SMILES
[Zr].CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O
Appearance (Color)
Colorless to pale yellow
Form
clear liquid
Assay (unspecified)
> 97.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text