Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, Thermo Scientific Chemicals

Catalog number: 042595.14
25 g, Each
Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, Thermo Scientific Chemicals

Catalog number: 042595.14
25 g, Each
Quantity
Catalog number: 042595.14
also known as 042595-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4039-32-1
IUPAC Name
lithium(1+) bis(trimethylsilyl)azanide
Molecular Formula
C6H18LiNSi2
InChI Key
YNESATAKKCNGOF-UHFFFAOYSA-N
SMILES
[Li+].C[Si](C)(C)[N-][Si](C)(C)C
Assay from Suppliers CofA
≥98.0 to ≤102.0% (Titration)
Form
Crystalline powder and/or chunks
Appearance (Color)
White to yellow

Description

Lithium bis(trimethylsilyl)amide is a base used in preparation of dienes and enolates. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. Lithium bis(trimethylsilyl)amide is also used in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text