Thermo Scientific Chemicals
Fullerene powder, 99+% C{70}, Thermo Scientific Chemicals
Catalog number: 042600.03
1 g, Each
Thermo Scientific Chemicals
Fullerene powder, 99+% C{70}, Thermo Scientific Chemicals
Catalog number: 042600.03
1 g, Each
Quantity
Catalog number: 042600.03
also known as 042600-03
Price (USD)
Have Questions?
Description
Fullerene powder is used for biomedical applications including the design of high-performance magnetic resonance imaging (MRI) contrast agents, X-ray imaging contrast agents, photodynamic therapy and drug and gene delivery. It is used in superconductors, photoactive polymers, film material modifiers, abrasive agent for cutting and grinding applications.