Chemical Identifiers
CAS115383-22-7
IUPAC Namehexatriacontacyclo[11.3.54.0¹⁷,²².0²,²³.0¹⁴,¹⁸.0¹⁹,⁷⁰.0²⁰,³⁷.0²¹,³⁴.0⁸,¹⁶.0¹⁰,¹⁵.0⁹,⁶³.0³,⁷.0⁴,²⁵.0⁶,⁶².0⁵,⁴⁸.0²⁴,³³.0³¹,³⁵.0²⁷,³².0²⁶,⁴⁷.0²⁸,⁴⁶.0⁴⁵,⁵⁰.0⁴⁹,⁶¹.0⁵¹,⁵⁹.0⁴⁴,⁵².0⁶⁰,⁶⁴.0¹¹,⁶⁵.0¹²,⁶⁷.0⁵⁸,⁶⁶.0⁵⁶,⁶⁸.0³⁸,⁶⁹.0³⁹,⁵⁵.0³⁶,⁴⁰.0³⁰,⁴¹.0²⁹,⁴³.0⁴²,⁵⁴.0⁵³,⁵⁷]heptaconta-1,3,5(48),6,8(16),9,11(65),12(67),13(70),14,17(22),18,20,23,25,27(32),28,30(41),31(35),33,36,38(69),39,42,44(52),45(50),46,49(61),51(59),53(57),54,56(68),58(66),60(64),62-pentatriacontaene
Molecular FormulaC70
InChI KeyATLMFJTZZPOKLC-UHFFFAOYSA-N
SMILESC12=C3C4=C5C1=C1C6=C7C8=C9C%10=C%11C%12=C%13C%14=C%15C%16=C%17C%18=C%19C%20=C%21C%22=C%19C%19=C%23C(C%11=C%19C%18=C%12%16)=C9C9=C%11C(=C2C9=C18)C1=C(C2=C%21C8=C9C(=C4C4=C9C9=C%12C(=C%17C%20=C8%12)C%15=C8C%14=C(C%12=C6C5=C4C%12=C98)C7=C%10%13)C2=C31)C%22=C%23%11
View more