Thermo Scientific Chemicals
Polyethylene, low density, ∣<400 micron, Thermo Scientific Chemicals
Catalog number: 042607.22
100 g, Each
Thermo Scientific Chemicals
Polyethylene, low density, ∣<400 micron, Thermo Scientific Chemicals
Catalog number: 042607.22
100 g, Each
Quantity
Catalog number: 042607.22
Price (USD)
Quantity
-
Have Questions?
Description
Polyethylene is used in injection and blow molded toys, housewares, lids, injection molded car seats, Hot melt coating for paper, additive in cast moldings, candles, oil-based inks and hot melt adhesives. It is also used as an additive, lubricant in rubber processing, extrusion and calendaring aid for PVC and dispersing aid for color concentrates. Bearings, gears, bushings and other moving parts.