Titanium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Titanium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042624.22
also known as 042624-22
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
Add to cart
Titanium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number042624.22
Price (USD)/ Each
77.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
91.30 
Save 13.65 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-325 mesh (less than 45 micron)
Trace Metalsless than 0.5% trace metal impurities
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only