Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%, Thermo Scientific Chemicals
Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%, Thermo Scientific Chemicals

Ion-pair reagent for HPLC analysis
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042636.06
also known as 042636-06
Price (USD)/ Each
44.65
Online exclusive
49.70 
Save 5.05 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
44.65
Online exclusive
49.70 
Save 5.05 (10%)
Add to cart
Sodium 1-octanesulfonate hydrate, HPLC grade, 99+%, Thermo Scientific Chemicals
Catalog number042636.06
Price (USD)/ Each
44.65
Online exclusive
49.70 
Save 5.05 (10%)
-
Add to cart
Chemical Identifiers
CAS5324-84-5
IUPAC Namesodium octane-1-sulfonate
Molecular FormulaC8H17NaO3S
InChI KeyHRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥99.0%
Water Content (Karl Fischer Titration)7.0-9.0%
Ion-pair reagent for HPLC analysisSodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only