Thermo Scientific Chemicals

Molybdenite, naturally occurring mineral, grains, ≈0.06 to 0.19 in., Thermo Scientific Chemicals

Catalog number: 042642.06
5 g, Each
Thermo Scientific Chemicals

Molybdenite, naturally occurring mineral, grains, ≈0.06 to 0.19 in., Thermo Scientific Chemicals

Catalog number: 042642.06
5 g, Each
Quantity
Catalog number: 042642.06
also known as 042642-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-56-4
IUPAC Name
disulfanylidenemolybdenum
Molecular Formula
MoS2
InChI Key
CWQXQMHSOZUFJS-UHFFFAOYSA-N
SMILES
S=[Mo]=S
Comment
Molybdenite is naturally occurring Molybdenum disulfide mined and crushed to approximately 0.06-0.19in without any chemical processing.

Description

Molybdenite, naturally occurring mineral, grains, is used for grain mounts, XRD and microprobe standards, as well as identification of unknown minerals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text