Thermo Scientific Chemicals

Iodine monochloride, approx. 0.22N soln. in glacial acetic acid, Thermo Scientific Chemicals

Catalog number: 042647.K2
1 L, Each
Thermo Scientific Chemicals

Iodine monochloride, approx. 0.22N soln. in glacial acetic acid, Thermo Scientific Chemicals

Catalog number: 042647.K2
1 L, Each
Quantity
Catalog number: 042647.K2
also known as 042647-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-99-0
IUPAC Name
iodochlorane
Molecular Formula
ClI
InChI Key
QZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILES
ClI
Form
Liquid
Appearance (Color)
Brown
Assay from Supplier's CofA
ca. 0.22N
Assay from Supplier's CofA
Further Ingredients: Acetic acid CAS 64-19-7, EEC No 200-580-7

Description

For determination of iodine absorption number of fatsIodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture and Light Sensitive
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text