Cobalt foil, 1.0mm (0.04in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Cobalt foil, 1.0mm (0.04in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt foil, 1.0mm (0.04in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number 042660.FI
also known as 042660-FI
Price (USD)/ Each
295.00
-
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)/ Each
295.00
Add to cart
Cobalt foil, 1.0mm (0.04in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number042660.FI
Price (USD)/ Each
295.00
-
Add to cart
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue silver
FormFoil, 1.0mm (0.04in) thick.
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only