Gold powder, spherical, APS 3.0-5.5 micron, 99.96+% (metals basis), Thermo Scientific Chemicals
Gold powder, spherical, APS 3.0-5.5 micron, 99.96+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold powder, spherical, APS 3.0-5.5 micron, 99.96+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 042674.06
also known as 042674-06
Price (USD)/ Each
1,312.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
1,312.00
Add to cart
Gold powder, spherical, APS 3.0-5.5 micron, 99.96+% (metals basis), Thermo Scientific Chemicals
Catalog number042674.06
Price (USD)/ Each
1,312.00
-
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Gold
Total Metal Impurities<0.04%
Particle Size3.0 - 5.5 microns
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only