Thermo Scientific Chemicals

Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals

Catalog number: 042679.K7
4 L, Each
Thermo Scientific Chemicals

Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals

Catalog number: 042679.K7
4 L, Each
Quantity
Catalog number: 042679.K7
also known as 042679-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7447-40-7
IUPAC Name
potassium chloride
Molecular Formula
ClK
InChI Key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES
[Cl-].[K+]
Conductivity
1413micromho at 25?C
Concentration
0.010M

Description

Potassium chloride 0.010M solution is used as conductive standard and as electrolyte in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text