Titanium(IV) oxide, rutile, 99.8% (metals basis), Thermo Scientific Chemicals
Titanium(IV) oxide, rutile, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Titanium(IV) oxide, rutile, 99.8% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number 042681.A1
also known as 042681-A1
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
Add to cart
Titanium(IV) oxide, rutile, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number042681.A1
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1317-80-2
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Titanium(IV) oxide is used in paints, plastics, paper, and foods. It is used as a UV-resistant material. It shows useful applications in earth sciences. It is also useful in quantitative provenance studies. It is also useful for the photocatalytic applications. It is also useful in high temperature superconducting resonators. It is also used as a material in the memristor, a new electronic circuit element. It can be used for solar energy conversion based on dye, to cleave protein that contains the amino acid proline polymer, quantum dot sensitized nanocrystalline TiO2 solar cells using conjugated polymers as solid electrolytes.