Thermo Scientific Chemicals

Tri-n-butylgermanium hydride, 97+%, Thermo Scientific Chemicals

Catalog number: 042688.03
1 g, Each
Thermo Scientific Chemicals

Tri-n-butylgermanium hydride, 97+%, Thermo Scientific Chemicals

Catalog number: 042688.03
1 g, Each
Quantity
Catalog number: 042688.03
also known as 042688-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
998-39-0
IUPAC Name
tributylgermyl
Molecular Formula
C12H27Ge
InChI Key
UMRPCNGKANTZSN-UHFFFAOYSA-N
SMILES
CCCC[Ge](CCCC)CCCC
Appearance (Color)
Clear, colorless to pale yellow
Form
Liquid
Assay from Supplier's CofA
≥97%
Identification (FTIR)
Conforms

Description

Tri-n-butylgermanium hydride is used in the reduction of benzylic chlorides. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text