Potassium chloride, 0.0375M, Conductivity Standard, Thermo Scientific Chemicals
Potassium chloride, 0.0375M, Conductivity Standard, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium chloride, 0.0375M, Conductivity Standard, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
4 L
Catalog number 042694.K2
also known as 042694-K2
Price (USD)/ Each
43.65
Online exclusive
48.10 
Save 4.45 (9%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
48.10 
Save 4.45 (9%)
Add to cart
Potassium chloride, 0.0375M, Conductivity Standard, Thermo Scientific Chemicals
Catalog number042694.K2
Price (USD)/ Each
43.65
Online exclusive
48.10 
Save 4.45 (9%)
-
Add to cart
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Formliquid
Conductivity4950 to 5050 micromhos at 25 deg C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only